Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTCAGTTCCCACAGCTTCTCTC[A/G]ATCATCCTCAGCCTATATCCAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147670 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INSR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INSR - insulin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000208.3 | 8308 | UTR 3 | NP_000199.2 | |||
NM_001079817.2 | 8308 | UTR 3 | NP_001073285.1 | |||
XM_011527988.2 | 8308 | UTR 3 | XP_011526290.2 | |||
XM_011527989.2 | 8308 | UTR 3 | XP_011526291.2 |