Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610837 MIM: 608846 MIM: 602950 | ||||||||||||||||||||
Literature Links: |
ADM5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADM5 - adrenomedullin 5 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BCL2L12 - BCL2 like 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CPT1C - carnitine palmitoyltransferase 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5088 - microRNA 5088 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRMT1 - protein arginine methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207042.2 | 342 | Silent Mutation | ACG,ACT | T,T 57 | NP_001193971.1 | |
NM_001536.5 | 342 | Silent Mutation | ACG,ACT | T,T 75 | NP_001527.3 | |
NM_198318.4 | 342 | Silent Mutation | ACG,ACT | T,T 57 | NP_938074.2 | |
XM_005258842.1 | 342 | Silent Mutation | ACG,ACT | T,T 46 | XP_005258899.1 | |
XM_017026734.1 | 342 | Silent Mutation | ACG,ACT | T,T 51 | XP_016882223.1 | |
XM_017026735.1 | 342 | Silent Mutation | ACG,ACT | T,T 51 | XP_016882224.1 | |
XM_017026736.1 | 342 | Silent Mutation | ACG,ACT | T,T 46 | XP_016882225.1 |