Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGCACTGTTGGTGCGGAAGCTT[C/T]GGGAGAAGCGGCAGACGGAGGGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609737 MIM: 613634 MIM: 608746 | ||||||||||||||||||||
Literature Links: |
CRB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRB3 - crumbs 3, cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139161.4 | 469 | Missense Mutation | CGG,TGG | R,W 84 | NP_631900.1 | |
NM_174881.3 | 469 | Missense Mutation | CGG,TGG | R,W 84 | NP_777377.1 | |
NM_174882.2 | 469 | Missense Mutation | CGG,TGG | R,W 84 | NP_777378.1 | |
XM_017027471.1 | 469 | Missense Mutation | CGG,TGG | R,W 147 | XP_016882960.1 |
DENND1C - DENN domain containing 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290331.1 | 469 | Intron | NP_001277260.1 | |||
NM_024898.3 | 469 | Intron | NP_079174.2 | |||
XM_006722905.2 | 469 | Intron | XP_006722968.2 | |||
XM_006722906.3 | 469 | Intron | XP_006722969.1 | |||
XM_011528318.2 | 469 | Intron | XP_011526620.1 |
SLC25A23 - solute carrier family 25 member 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |