Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTACAAACCATCACCAGCACCAG[A/C]AGCATCACAGTGCAAAGAAATCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZIK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZIK1 - zinc finger protein interacting with K protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010879.3 | 751 | Missense Mutation | AAG,CAG | K,Q 137 | NP_001010879.2 | |
NM_001321145.1 | 751 | Missense Mutation | AAG,CAG | K,Q 124 | NP_001308074.1 | |
NM_001321146.1 | 751 | Missense Mutation | AAG,CAG | K,Q 82 | NP_001308075.1 | |
NM_001321147.1 | 751 | Missense Mutation | AAG,CAG | K,Q 34 | NP_001308076.1 | |
XM_011526761.1 | 751 | Missense Mutation | AAG,CAG | K,Q 123 | XP_011525063.1 | |
XM_011526762.1 | 751 | Missense Mutation | AAG,CAG | K,Q 118 | XP_011525064.1 | |
XM_011526763.1 | 751 | Missense Mutation | AAG,CAG | K,Q 110 | XP_011525065.1 | |
XM_011526764.1 | 751 | Missense Mutation | AAG,CAG | K,Q 104 | XP_011525066.1 | |
XM_011526765.2 | 751 | Missense Mutation | AAG,CAG | K,Q 101 | XP_011525067.1 | |
XM_011526766.2 | 751 | Missense Mutation | AAG,CAG | K,Q 82 | XP_011525068.1 | |
XM_011526767.2 | 751 | Missense Mutation | AAG,CAG | K,Q 34 | XP_011525069.1 |
ZNF530 - zinc finger protein 530 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |