Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTCGAAGGTTGGGCGAAAGGAC[A/G]CCTCTGTCTCCCAGCAGTTCTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176941 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TYK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TYK2 - tyrosine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003331.4 | 3580 | Missense Mutation | GCG,GTG | A,V 1156 | NP_003322.3 | |
XM_011528245.1 | 3580 | Missense Mutation | GCG,GTG | A,V 1156 | XP_011526547.1 | |
XM_011528246.2 | 3580 | Missense Mutation | GCG,GTG | A,V 1057 | XP_011526548.1 | |
XM_011528247.1 | 3580 | Missense Mutation | GCG,GTG | A,V 1057 | XP_011526549.1 | |
XM_011528249.2 | 3580 | Missense Mutation | GCG,GTG | A,V 714 | XP_011526551.1 | |
XM_011528251.1 | 3580 | Missense Mutation | GCG,GTG | A,V 575 | XP_011526553.1 | |
XM_011528252.1 | 3580 | Intron | XP_011526554.1 |