Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCCAGGTTTTTCCGCCAACTAG[A/G]CCGGGACGGGAGCAGATCCCTGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114212 MIM: 603327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPS - calcyphosine | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004058.4 | 789 | Missense Mutation | GAC,GGC | D,G 120 | NP_004049.2 | |
NM_080590.3 | 789 | Missense Mutation | GAC,GGC | D,G 120 | NP_542157.2 |
RANBP3 - RAN binding protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VMAC - vimentin-type intermediate filament associated coiled-coil protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |