Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGACAGACGGATCCTGACCCTCA[A/C]ACTGGAGTGGGTCACAAGGGCGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609094 MIM: 603021 | ||||||||||||||||||||
Literature Links: |
FBXO17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO17 - F-box protein 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024907.6 | 943 | Missense Mutation | GTG,TTG | V,L 271 | NP_079183.4 | |
NM_148169.2 | 943 | Missense Mutation | GTG,TTG | V,L 280 | NP_680474.1 |
MRPS12 - mitochondrial ribosomal protein S12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |