Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGCTGAGTTACGAGCAGAGCAA[A/G]CATAGAGCCCATTATGCTTTGGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176401 | ||||||||||||||||||||
Literature Links: |
PSG11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSG11 - pregnancy specific beta-1-glycoprotein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113410.1 | 640 | Missense Mutation | GCT,GTT | A,V 178 | NP_001106881.1 | |
NM_002785.2 | 640 | Missense Mutation | GCT,GTT | A,V 300 | NP_002776.3 | |
NM_203287.1 | 640 | Missense Mutation | GCT,GTT | A,V 178 | NP_976032.2 |