Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTGCCCTGTTGGGTGATAACGT[A/C]CACAGGTAGTGGGGGACCTCTTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF234 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF234 - zinc finger protein 234 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144824.1 | Intron | NP_001138296.1 | ||||
NM_006630.2 | Intron | NP_006621.1 | ||||
XM_006722974.3 | Intron | XP_006723037.1 | ||||
XM_017026149.1 | Intron | XP_016881638.1 | ||||
XM_017026150.1 | Intron | XP_016881639.1 | ||||
XM_017026151.1 | Intron | XP_016881640.1 | ||||
XM_017026152.1 | Intron | XP_016881641.1 | ||||
XM_017026153.1 | Intron | XP_016881642.1 |