Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTGGCTTGGTCTCCGCTTCAAC[C/T]GGCTGCTAGGGAAGATGCTAAAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 151440 MIM: 164005 MIM: 611669 | ||||||||||||||||||||
Literature Links: |
LYL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LYL1 - LYL1, basic helix-loop-helix family member | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005583.4 | 1590 | Missense Mutation | CAG,CGG | Q,R 128 | NP_005574.2 | |
XM_005259912.3 | 1590 | Missense Mutation | CAG,CGG | Q,R 54 | XP_005259969.1 | |
XM_017026816.1 | 1590 | Missense Mutation | CAG,CGG | Q,R 199 | XP_016882305.1 | |
XM_017026817.1 | 1590 | Missense Mutation | CAG,CGG | Q,R 150 | XP_016882306.1 |
NFIX - nuclear factor I X | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT1 - tRNA methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |