Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCTCCTCCTCACAGGACTCTCC[A/C]CCTCTCGACCTCGTGTGGTCATCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF324 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF324 - zinc finger protein 324 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014347.2 | 1070 | Missense Mutation | ACC,CCC | T,P 44 | NP_055162.1 | |
XM_005258713.4 | 1070 | Missense Mutation | ACC,CCC | T,P 49 | XP_005258770.1 |
ZNF446 - zinc finger protein 446 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |