Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGCCGTCGCCGGCATCGTGAC[C/T]GGGGTCCTGGTCGGAGTGGCGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609142 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEACAM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEACAM3 - carcinoembryonic antigen related cell adhesion molecule 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277163.2 | 562 | Silent Mutation | ACC,ACT | T,T 158 | NP_001264092.1 | |
NM_001815.4 | 562 | Silent Mutation | ACC,ACT | T,T 158 | NP_001806.2 |