Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTGCGCACTTTGGGCTTGGCCA[A/G]GGAGATGATCCGGGGGCTGGCCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609503 | ||||||||||||||||||||
Literature Links: |
THEG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
THEG - theg spermatid protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016585.4 | 1760 | Silent Mutation | CTG,TTG | L,L 339 | NP_057669.1 | |
NM_199202.2 | 1760 | Silent Mutation | CTG,TTG | L,L 315 | NP_954672.1 | |
XM_011528049.2 | 1760 | Silent Mutation | CTG,TTG | L,L 407 | XP_011526351.1 | |
XM_011528050.2 | 1760 | Silent Mutation | CTG,TTG | L,L 383 | XP_011526352.1 | |
XM_011528051.2 | 1760 | Silent Mutation | CTG,TTG | L,L 339 | XP_011526353.1 | |
XM_011528052.2 | 1760 | Intron | XP_011526354.1 |