Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTGATTCTCCCGGGATACACACC[A/G]TGGCAGGTGGCATGGGAGACGATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604434 MIM: 605539 | ||||||||||||||||||||
Literature Links: |
KLK11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLK11 - kallikrein related peptidase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLK12 - kallikrein related peptidase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019598.2 | 376 | Silent Mutation | CAC,CAT | H,H 173 | NP_062544.1 | |
NM_145894.1 | 376 | Silent Mutation | CAC,CAT | H,H 173 | NP_665901.1 | |
NM_145895.1 | 376 | Missense Mutation | CGG,TGG | R,W 87 | NP_665902.1 | |
XM_005258950.3 | 376 | Silent Mutation | CAC,CAT | H,H 63 | XP_005259007.1 | |
XM_005258951.3 | 376 | Missense Mutation | CGG,TGG | R,W 87 | XP_005259008.1 |