Search Thermo Fisher Scientific
GGCTTCTCGGCCGGCACTGAGTACA[C/T]GGACCTGCAGAATGACCTAGGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604495 MIM: 604480 | ||||||||||||||||||||
Literature Links: |
CCER2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCER2 - coiled-coil glutamate rich protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFKBIB - NFKB inhibitor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243116.1 | 299 | Missense Mutation | ACG,ATG | T,M 1 | NP_001230045.1 | |
NM_002503.4 | 299 | Missense Mutation | ACG,ATG | T,M 87 | NP_002494.2 | |
XM_006723226.2 | 299 | Missense Mutation | ACG,ATG | T,M 55 | XP_006723289.2 | |
XM_006723227.3 | 299 | Missense Mutation | ACG,ATG | T,M 82 | XP_006723290.3 |
SIRT2 - sirtuin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |