Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATCTTCACCTCGCTGGAGTACG[G/T]ACCGGTGCCGGAGAGCCACGCATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613358 MIM: 611480 | ||||||||||||||||||||
Literature Links: |
ALDH16A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH16A1 - aldehyde dehydrogenase 16 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145396.1 | 175 | Missense Mutation | GGA,GTA | G,V 20 | NP_001138868.1 | |
NM_153329.3 | 175 | Missense Mutation | GGA,GTA | G,V 20 | NP_699160.2 | |
XM_011526441.1 | 175 | Intron | XP_011524743.1 | |||
XM_011526442.1 | 175 | Intron | XP_011524744.1 |
PIH1D1 - PIH1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017916.2 | 175 | Intron | NP_060386.1 |