Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCCCCCGCTCCCGAAAACCCGG[C/T]GCCGTCATCGAGAGCTTTGTGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606700 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIDN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIDN - midnolin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177401.4 | 1442 | Silent Mutation | GGC,GGT | G,G 266 | NP_796375.3 | |
XM_005259671.3 | 1442 | Silent Mutation | GGC,GGT | G,G 309 | XP_005259728.1 | |
XM_005259672.3 | 1442 | Silent Mutation | GGC,GGT | G,G 308 | XP_005259729.1 |