Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCATGGAACAGAGGATGAAGAG[A/C]CACCTTCTGACCAGAATGTTTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZIK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZIK1 - zinc finger protein interacting with K protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010879.3 | 571 | Missense Mutation | ACA,CCA | T,P 77 | NP_001010879.2 | |
NM_001321145.1 | 571 | Missense Mutation | ACA,CCA | T,P 64 | NP_001308074.1 | |
NM_001321146.1 | 571 | Missense Mutation | ACA,CCA | T,P 22 | NP_001308075.1 | |
NM_001321147.1 | 571 | UTR 5 | NP_001308076.1 | |||
XM_011526761.1 | 571 | Missense Mutation | ACA,CCA | T,P 63 | XP_011525063.1 | |
XM_011526762.1 | 571 | Missense Mutation | ACA,CCA | T,P 58 | XP_011525064.1 | |
XM_011526763.1 | 571 | Missense Mutation | ACA,CCA | T,P 50 | XP_011525065.1 | |
XM_011526764.1 | 571 | Missense Mutation | ACA,CCA | T,P 44 | XP_011525066.1 | |
XM_011526765.2 | 571 | Missense Mutation | ACA,CCA | T,P 41 | XP_011525067.1 | |
XM_011526766.2 | 571 | Missense Mutation | ACA,CCA | T,P 22 | XP_011525068.1 | |
XM_011526767.2 | 571 | UTR 5 | XP_011525069.1 |
ZNF530 - zinc finger protein 530 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |