Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTCATCTCCAGCGACTTCACAG[A/C]CGTGCTGCAGACGCTGCCTCGCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606377 | ||||||||||||||||||||
Literature Links: |
DHDH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHDH - dihydrodiol dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014475.3 | 96 | Missense Mutation | GAC,GCC | D,A 19 | NP_055290.1 | |
XM_005258748.4 | 96 | Intron | XP_005258805.1 | |||
XM_017026598.1 | 96 | Intron | XP_016882087.1 |