Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCTGGCGGCTAAACCGTGGTTT[C/G]AGCCAGGCTGCCCTCCTGCTGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608348 MIM: 606492 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCKDHA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCKDHA - branched chain keto acid dehydrogenase E1, alpha polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000709.3 | 90 | Missense Mutation | TTC,TTG | F,L 17 | NP_000700.1 | |
NM_001164783.1 | 90 | Missense Mutation | TTC,TTG | F,L 17 | NP_001158255.1 |
EXOSC5 - exosome component 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020158.3 | 90 | Intron | NP_064543.3 |