Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTTATGTGTCTCAAGGGTTGAT[A/C]CACAACTGAAAACCTTTTCACATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF600 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF600 - zinc finger protein 600 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321866.1 | Intron | NP_001308795.1 | ||||
NM_001321867.1 | Intron | NP_001308796.1 | ||||
NM_198457.3 | Intron | NP_940859.3 | ||||
XM_017026390.1 | Intron | XP_016881879.1 | ||||
XM_017026391.1 | Intron | XP_016881880.1 | ||||
XM_017026392.1 | Intron | XP_016881881.1 |