Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTCCTGTGTCTTTCTCTCCAGA[A/T]TCACAGCTCGGAGGAAGGTCTCCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601270 | ||||||||||||||||||||
Literature Links: |
CYP4F3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP4F3 - cytochrome P450 family 4 subfamily F member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000896.2 | 2 | Missense Mutation | AAT,ATT | N,I 67 | NP_000887.2 | |
NM_001199208.1 | 2 | Intron | NP_001186137.1 | |||
NM_001199209.1 | 2 | Intron | NP_001186138.1 | |||
XM_005259911.4 | 2 | Intron | XP_005259968.1 | |||
XM_011528014.2 | 2 | UTR 5 | XP_011526316.1 | |||
XM_017026814.1 | 2 | Intron | XP_016882303.1 | |||
XM_017026815.1 | 2 | Intron | XP_016882304.1 |