Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTCCGACTCCTCATCCCGCACA[A/G]GCATCTGGTAGGACCTGAGGGAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614987 MIM: 613245 | ||||||||||||||||||||
Literature Links: |
EPS8L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPS8L1 - EPS8 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R12C - protein phosphatase 1 regulatory subunit 12C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271618.1 | 1653 | Missense Mutation | NP_001258547.1 | |||
NM_017607.3 | 1653 | Missense Mutation | NP_060077.1 | |||
XM_005259013.3 | 1653 | Missense Mutation | XP_005259070.1 | |||
XM_011527045.2 | 1653 | Missense Mutation | XP_011525347.1 |