Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTCACATTTATATGGTTTCTCTC[C/T]GGTGTGGACTCTCTGATGGGCTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604749 | ||||||||||||||||||||
Literature Links: |
ZNF235 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF235 - zinc finger protein 235 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004234.4 | 6026 | Missense Mutation | AGA,GGA | R,G 623 | NP_004225.3 | |
XM_005259412.4 | 6026 | Missense Mutation | AGA,GGA | R,G 619 | XP_005259469.1 |