Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATTATTCCCATAGTTATTAGAAA[A/T]ACGGGTTTTGGGACTACAAGAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF321P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF321P - zinc finger protein 321, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF816-ZNF321P - ZNF816-ZNF321P readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202473.1 | Intron | NP_001189402.1 |
ZNF888 - zinc finger protein 888 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310127.1 | Intron | NP_001297056.1 | ||||
XM_005259451.4 | Intron | XP_005259508.1 | ||||
XM_017026798.1 | Intron | XP_016882287.1 | ||||
XM_017026799.1 | Intron | XP_016882288.1 | ||||
XM_017026800.1 | Intron | XP_016882289.1 |