Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACCGGGTTTACCCAAACTCAAC[A/G]TGCAAGCCTCGGATTATTGCACCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123837 | ||||||||||||||||||||
Literature Links: |
CCNE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCNE1 - cyclin E1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001238.3 | 456 | Silent Mutation | ACA,ACG | T,T 91 | NP_001229.1 | |
NM_001322259.1 | 456 | Silent Mutation | ACA,ACG | T,T 91 | NP_001309188.1 | |
NM_001322261.1 | 456 | Silent Mutation | ACA,ACG | T,T 91 | NP_001309190.1 | |
NM_001322262.1 | 456 | Silent Mutation | ACA,ACG | T,T 76 | NP_001309191.1 | |
XM_011527440.2 | 456 | Silent Mutation | ACA,ACG | T,T 88 | XP_011525742.1 |