Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCAGGCCCTGACTGCACACTCCC[A/G]TTTTGGCAGGAAACTAAAATTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602378 MIM: 605395 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf38 - chromosome 19 open reading frame 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DNM2 - dynamin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005360.2 | 899 | Intron | NP_001005360.1 | |||
NM_001005361.2 | 899 | Intron | NP_001005361.1 | |||
NM_001005362.2 | 899 | Intron | NP_001005362.1 | |||
NM_001190716.1 | 899 | Intron | NP_001177645.1 | |||
NM_004945.3 | 899 | Intron | NP_004936.2 |
MIR6793 - microRNA 6793 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMED1 - transmembrane p24 trafficking protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006858.3 | 899 | UTR 3 | NP_006849.1 | |||
XM_006722631.3 | 899 | UTR 3 | XP_006722694.1 |