Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTTTTGCAGGCTGCCTCACTCA[C/G]ATATTTTTTTTCATTGCATTTGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600637 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR7C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR7C2 - olfactory receptor family 7 subfamily C member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012377.1 | 300 | Missense Mutation | CAC,CAG | H,Q 100 | NP_036509.1 |
SLC1A6 - solute carrier family 1 member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |