Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCGGATTGTGGGAGGCTGGGAGT[A/G]TGAGAAGCATTCCCAACCCTGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147960 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLK2 - kallikrein related peptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002231.2 | 133 | Missense Mutation | TAT,TGT | Y,C 31 | NP_001002231.1 | |
NM_001256080.1 | 133 | Intron | NP_001243009.1 | |||
NM_005551.4 | 133 | Missense Mutation | TAT,TGT | Y,C 31 | NP_005542.1 |
KLKP1 - kallikrein pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |