Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCTATAATGGATTAAGGCCAG[A/G]AAGATGTCCTGTTCCTGAAGGAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176392 | ||||||||||||||||||||
Literature Links: |
PSG3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSG3 - pregnancy specific beta-1-glycoprotein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021016.3 | 1399 | Missense Mutation | CCT,TCT | P,S 423 | NP_066296.2 | |
XM_011527126.1 | 1399 | Missense Mutation | CCT,TCT | P,S 352 | XP_011525428.1 | |
XM_011527127.1 | 1399 | Missense Mutation | CCT,TCT | P,S 267 | XP_011525429.1 |