Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGCTGGACTTTGTCTAGGGAGAT[G/T]CGGAGCAGGGCTTGCTGGTAGCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600941 MIM: 612125 | ||||||||||||||||||||
Literature Links: |
BLVRB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BLVRB - biliverdin reductase B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105372489 - uncharacterized LOC105372489 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERTAD3 - SERTA domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013368.3 | 383 | Silent Mutation | CGA,CGC | R,R 35 | NP_037500.2 | |
NM_203344.2 | 383 | Silent Mutation | CGA,CGC | R,R 35 | NP_976219.1 | |
XM_005258832.3 | 383 | Silent Mutation | CGA,CGC | R,R 35 | XP_005258889.1 | |
XM_006723179.3 | 383 | Silent Mutation | CGA,CGC | R,R 35 | XP_006723242.1 |