Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCCACAAACAGGCAAGCCACGCC[C/T]AGCAGCTGCAGGCGATGTAGACGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164731 MIM: 600137 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKT2 - AKT serine/threonine kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CNTD2 - cyclin N-terminal domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024877.3 | 508 | Silent Mutation | CTA,CTG | L,L 153 | NP_079153.2 | |
XM_006723395.3 | 508 | Silent Mutation | CTA,CTG | L,L 221 | XP_006723458.2 |
MAP3K10 - mitogen-activated protein kinase kinase kinase 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC9B - tetratricopeptide repeat domain 9B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |