Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGATGAGGTCCTCGGGACACCC[C/T]GTCTTCTTGGAGCCCCCCAGGCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601670 MIM: 616655 | ||||||||||||||||||||
Literature Links: |
DPF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPF1 - double PHD fingers 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135155.2 | 909 | Silent Mutation | ACA,ACG | T,T 311 | NP_001128627.1 | |
NM_001135156.2 | 909 | Silent Mutation | ACA,ACG | T,T 229 | NP_001128628.1 | |
NM_001289978.1 | 909 | Silent Mutation | ACA,ACG | T,T 311 | NP_001276907.1 | |
NM_004647.3 | 909 | Silent Mutation | ACA,ACG | T,T 267 | NP_004638.2 | |
XM_005259289.2 | 909 | Silent Mutation | ACA,ACG | T,T 294 | XP_005259346.1 | |
XM_005259290.3 | 909 | Silent Mutation | ACA,ACG | T,T 285 | XP_005259347.1 | |
XM_005259292.4 | 909 | Silent Mutation | ACA,ACG | T,T 266 | XP_005259349.1 | |
XM_006723407.2 | 909 | Silent Mutation | ACA,ACG | T,T 285 | XP_006723470.1 | |
XM_006723408.3 | 909 | Silent Mutation | ACA,ACG | T,T 283 | XP_006723471.1 | |
XM_006723409.2 | 909 | Silent Mutation | ACA,ACG | T,T 129 | XP_006723472.1 | |
XM_006723410.2 | 909 | Silent Mutation | ACA,ACG | T,T 129 | XP_006723473.1 | |
XM_011527356.1 | 909 | Silent Mutation | ACA,ACG | T,T 294 | XP_011525658.1 | |
XM_011527357.2 | 909 | Silent Mutation | ACA,ACG | T,T 266 | XP_011525659.1 | |
XM_011527358.2 | 909 | Silent Mutation | ACA,ACG | T,T 229 | XP_011525660.1 |
SIPA1L3 - signal induced proliferation associated 1 like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |