Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAGCCTCACCATGGAGTCCCCA[A/G]TGTAGCGTGTGAGGCACAGGAACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604996 MIM: 608303 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FXYD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FXYD3 - FXYD domain containing ion transport regulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LGI4 - leucine rich repeat LGI family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139284.2 | 1281 | Missense Mutation | ACT,ATT | T,I 429 | NP_644813.1 | |
XM_011526595.1 | 1281 | Missense Mutation | ACT,ATT | T,I 257 | XP_011524897.1 | |
XM_017026428.1 | 1281 | Missense Mutation | ACT,ATT | T,I 257 | XP_016881917.1 | |
XM_017026429.1 | 1281 | Missense Mutation | ACT,ATT | T,I 257 | XP_016881918.1 | |
XM_017026430.1 | 1281 | Missense Mutation | ACT,ATT | T,I 257 | XP_016881919.1 |
MIR6887 - microRNA 6887 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |