Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCCAATAACTTGAAGGCCCGCA[A/G]TTCCTTCCGCTACAACGGACTGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603638 | ||||||||||||||||||||
Literature Links: |
MIR6805 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6805 - microRNA 6805 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL28 - ribosomal protein L28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000991.4 | 149 | Missense Mutation | AAT,AGT | N,S 36 | NP_000982.2 | |
NM_001136134.1 | 149 | Missense Mutation | AAT,AGT | N,S 36 | NP_001129606.1 | |
NM_001136135.1 | 149 | Missense Mutation | AAT,AGT | N,S 36 | NP_001129607.1 | |
NM_001136136.1 | 149 | Missense Mutation | AAT,AGT | N,S 36 | NP_001129608.1 | |
NM_001136137.1 | 149 | Missense Mutation | AAT,AGT | N,S 36 | NP_001129609.1 |
TMEM190 - transmembrane protein 190 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM238 - transmembrane protein 238 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |