Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAGAGGGTCTGTCTGGCTCCCCA[A/G]TGAGCTTCCAGACCAAGCCCAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608258 MIM: 606746 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPP9 - dipeptidyl peptidase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139159.4 | Intron | NP_631898.3 | ||||
XM_005259673.2 | Intron | XP_005259730.1 | ||||
XM_011528405.1 | Intron | XP_011526707.1 | ||||
XM_011528406.1 | Intron | XP_011526708.1 | ||||
XM_011528408.1 | Intron | XP_011526710.1 | ||||
XM_011528410.1 | Intron | XP_011526712.1 | ||||
XM_017027453.1 | Intron | XP_016882942.1 | ||||
XM_017027454.1 | Intron | XP_016882943.1 |
DPP9-AS1 - DPP9 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYDGF - myeloid derived growth factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |