Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCACTAATGCCCAGCTGCTCCCC[A/G]GAGGTCTCTGTCTCTGTGGGCTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610221 MIM: 605610 MIM: 616659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKT1S1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKT1S1 - AKT1 substrate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098632.2 | 731 | Silent Mutation | TCC,TCT | S,S 116 | NP_001092102.1 | |
NM_001098633.3 | 731 | Silent Mutation | TCC,TCT | S,S 116 | NP_001092103.1 | |
NM_001278159.1 | 731 | Silent Mutation | TCC,TCT | S,S 116 | NP_001265088.1 | |
NM_001278160.1 | 731 | Silent Mutation | TCC,TCT | S,S 116 | NP_001265089.1 | |
NM_032375.5 | 731 | Silent Mutation | TCC,TCT | S,S 136 | NP_115751.3 |
PNKP - polynucleotide kinase 3'-phosphatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D17 - TBC1 domain family member 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |