Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCCTCCCTCTGGGCCCCGAGGAA[C/T]CCTCCTTCTGTTGCCGCTGCTGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601323 MIM: 602309 | ||||||||||||||||||||
Literature Links: |
NUCB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUCB1 - nucleobindin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006184.5 | 360 | Missense Mutation | ACC,ATC | T,I 9 | NP_006175.2 | |
XM_017026845.1 | 360 | Missense Mutation | ACC,ATC | T,I 9 | XP_016882334.1 |
TULP2 - tubby like protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003323.2 | 360 | Intron | NP_003314.2 | |||
XM_006723347.3 | 360 | Intron | XP_006723410.1 | |||
XM_006723348.3 | 360 | Intron | XP_006723411.1 | |||
XM_011527252.2 | 360 | Intron | XP_011525554.1 | |||
XM_011527253.2 | 360 | Intron | XP_011525555.1 | |||
XM_011527254.2 | 360 | Intron | XP_011525556.1 | |||
XM_011527255.2 | 360 | Intron | XP_011525557.1 | |||
XM_011527256.2 | 360 | Intron | XP_011525558.1 |