Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCCGGACGAGGAACTTGAGGAC[C/T]GTCCCCGCCTCAGAACCCAGGAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611289 MIM: 608873 | ||||||||||||||||||||
Literature Links: |
LRG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRG1 - leucine rich alpha-2-glycoprotein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA6B - semaphorin 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032108.3 | 1621 | Silent Mutation | NP_115484.2 | |||
XM_011527639.2 | 1621 | Silent Mutation | XP_011525941.1 | |||
XM_011527640.2 | 1621 | Silent Mutation | XP_011525942.1 | |||
XM_011527641.1 | 1621 | Silent Mutation | XP_011525943.1 |