Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCCAGAGAGCGTACACTGCCCC[A/G]CTGTTCATCAGCCCCATACTCTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607463 | ||||||||||||||||||||
Literature Links: |
PPP1R13L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1R13L - protein phosphatase 1 regulatory subunit 13 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142502.1 | 2475 | Silent Mutation | AGC,AGT | S,S 760 | NP_001135974.1 | |
NM_006663.3 | 2475 | Silent Mutation | AGC,AGT | S,S 760 | NP_006654.2 | |
XM_017026177.1 | 2475 | Silent Mutation | AGC,AGT | S,S 760 | XP_016881666.1 | |
XM_017026178.1 | 2475 | Silent Mutation | AGC,AGT | S,S 760 | XP_016881667.1 | |
XM_017026179.1 | 2475 | Intron | XP_016881668.1 |