Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTCACCCCCTTGTCGATGGTGTG[C/T]GTCTGGCACAGCGTGCCCTCGGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608990 | ||||||||||||||||||||
Literature Links: |
ADAMTS10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAMTS10 - ADAM metallopeptidase with thrombospondin type 1 motif 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282352.1 | 1875 | Intron | NP_001269281.1 | |||
NM_030957.3 | 1875 | Silent Mutation | ACA,ACG | T,T 523 | NP_112219.3 | |
XM_006722917.3 | 1875 | Silent Mutation | ACA,ACG | T,T 155 | XP_006722980.1 | |
XM_017027338.1 | 1875 | Silent Mutation | ACA,ACG | T,T 523 | XP_016882827.1 | |
XM_017027339.1 | 1875 | Silent Mutation | ACA,ACG | T,T 46 | XP_016882828.1 | |
XM_017027340.1 | 1875 | Silent Mutation | ACA,ACG | T,T 44 | XP_016882829.1 |