Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACATGCAGGAGACGCGGTTGCCG[C/T]GGGCGCTCTTGGTGGGGAAGACCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD17A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABHD17A - abhydrolase domain containing 17A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130111.1 | 3562 | Intron | NP_001123583.1 | |||
NM_031213.3 | 3562 | Intron | NP_112490.3 | |||
XM_011528338.1 | 3562 | Missense Mutation | CAC,CGC | H,R 95 | XP_011526640.1 | |
XM_017027341.1 | 3562 | Missense Mutation | CAC,CGC | H,R 95 | XP_016882830.1 | |
XM_017027342.1 | 3562 | UTR 5 | XP_016882831.1 |