Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGCTCCGGGAGATGGGAGATAA[C/G]TGACACTGAGACTTCATACCCAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610545 MIM: 610546 | ||||||||||||||||||||
Literature Links: |
IGFL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGFL2 - IGF like family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002915.2 | 2773 | Intron | NP_001002915.2 | |||
NM_001135113.1 | 2773 | Intron | NP_001128585.1 | |||
XM_011526501.1 | 2773 | Intron | XP_011524803.1 | |||
XM_017026335.1 | 2773 | Intron | XP_016881824.1 |
IGFL3 - IGF like family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207393.1 | 2773 | Missense Mutation | CAC,CAG | H,Q 107 | NP_997276.1 | |
XM_011526953.2 | 2773 | Missense Mutation | CAC,CAG | H,Q 107 | XP_011525255.1 | |
XM_017026796.1 | 2773 | Missense Mutation | CAC,CAG | H,Q 144 | XP_016882285.1 | |
XM_017026797.1 | 2773 | Missense Mutation | CAC,CAG | H,Q 144 | XP_016882286.1 |