Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAGGACCCTGGCGCGGCTCTCAC[A/G]GGCGTCATGGAGACCACGATGCAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613176 | ||||||||||||||||||||
Literature Links: |
IRGC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IRGC - immunity related GTPase cinema | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019612.3 | 442 | Silent Mutation | ACA,ACG | T,T 87 | NP_062558.1 | |
XM_005259058.3 | 442 | Silent Mutation | ACA,ACG | T,T 124 | XP_005259115.1 | |
XM_006723283.3 | 442 | Silent Mutation | ACA,ACG | T,T 87 | XP_006723346.1 | |
XM_011527118.2 | 442 | Silent Mutation | ACA,ACG | T,T 87 | XP_011525420.1 |
SMG9 - SMG9, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |