Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATAAGCTTTCTCCCCAGGATAC[A/G]TTTTTTGAGGTTGAGAGGATACTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF585A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF585A - zinc finger protein 585A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288800.1 | 627 | Missense Mutation | ACG,ATG | T,M 123 | NP_001275729.1 | |
NM_152655.3 | 627 | Missense Mutation | ACG,ATG | T,M 68 | NP_689868.1 | |
NM_199126.2 | 627 | Missense Mutation | ACG,ATG | T,M 68 | NP_954577.1 |