Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCACAATGGAGCCCCTGATGCTA[C/G]GTACAGCTTGAATACTTAATTTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF677 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF677 - zinc finger protein 677 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317998.1 | 2164 | Silent Mutation | ACC,ACG | T,T 584 | NP_001304927.1 | |
NM_182609.3 | 2164 | Silent Mutation | ACC,ACG | T,T 584 | NP_872415.1 | |
XM_005258862.4 | 2164 | Silent Mutation | ACC,ACG | T,T 548 | XP_005258919.1 | |
XM_011526912.2 | 2164 | Silent Mutation | ACC,ACG | T,T 584 | XP_011525214.1 |