Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCGGCTCCTGCACCACCAGCGCA[C/T]GCACACAGGCGAGCGGCCCTACAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610671 | ||||||||||||||||||||
Literature Links: |
NAT14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAT14 - N-acetyltransferase 14 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SSC5D - scavenger receptor cysteine rich family member with 5 domains | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF628 - zinc finger protein 628 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033113.2 | 368 | Missense Mutation | ACG,ATG | T,M 57 | NP_149104.3 | |
XM_005259371.3 | 368 | Missense Mutation | ACG,ATG | T,M 53 | XP_005259428.1 |