Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGCAATGTTGGTCAGATGTCAG[G/T]TTTGAGCGCTTCCTTCTGCACAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610095 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIR3DL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIR3DL3 - killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153443.4 | 497 | Missense Mutation | AGG,AGT | R,S 149 | NP_703144.3 |