Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCGGGGGCCTGGGACCAGCTTC[A/G]AGTTTGCGCTTGCAATTGTTGAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602533 | ||||||||||||||||||||
Literature Links: |
PARK7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PARK7 - Parkinsonism associated deglycase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123377.1 | Intron | NP_001116849.1 | ||||
NM_007262.4 | Intron | NP_009193.2 | ||||
XM_005263424.3 | Intron | XP_005263481.1 |