Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCGTTACCGTTCCGGGGCGGCC[C/G]CCTAGGCGGGTCCCGACGCGGCCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MORN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MORN1 - MORN repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301060.1 | 279 | Missense Mutation | CGG,GGG | R,G 21 | NP_001287989.1 | |
NM_024848.2 | 279 | Missense Mutation | CGG,GGG | R,G 21 | NP_079124.1 | |
XM_005244798.2 | 279 | Missense Mutation | CGG,GGG | R,G 21 | XP_005244855.1 | |
XM_011542169.1 | 279 | Missense Mutation | CGG,GGG | R,G 21 | XP_011540471.1 | |
XM_011542170.2 | 279 | Missense Mutation | CGG,GGG | R,G 21 | XP_011540472.1 | |
XM_011542172.2 | 279 | Missense Mutation | CGG,GGG | R,G 21 | XP_011540474.1 | |
XM_011542173.2 | 279 | Missense Mutation | CGG,GGG | R,G 21 | XP_011540475.1 | |
XM_011542174.2 | 279 | Missense Mutation | CGG,GGG | R,G 21 | XP_011540476.1 | |
XM_011542176.1 | 279 | Missense Mutation | CGG,GGG | R,G 21 | XP_011540478.1 | |
XM_011542177.1 | 279 | Missense Mutation | CGG,GGG | R,G 21 | XP_011540479.1 | |
XM_017002365.1 | 279 | Missense Mutation | CGG,GGG | R,G 21 | XP_016857854.1 |
RER1 - retention in endoplasmic reticulum sorting receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007033.4 | 279 | Intron | NP_008964.3 | |||
XM_005244713.4 | 279 | Intron | XP_005244770.2 | |||
XM_006710306.2 | 279 | Intron | XP_006710369.1 | |||
XM_011540542.1 | 279 | Intron | XP_011538844.1 | |||
XM_011540543.1 | 279 | Intron | XP_011538845.1 | |||
XM_017000131.1 | 279 | Intron | XP_016855620.1 | |||
XM_017000132.1 | 279 | Intron | XP_016855621.1 | |||
XM_017000133.1 | 279 | Intron | XP_016855622.1 | |||
XM_017000134.1 | 279 | Intron | XP_016855623.1 | |||
XM_017000135.1 | 279 | Intron | XP_016855624.1 |